Cintape® Self-Adhesive Indicator Tape for Hydrogen Peroxide (FDA)
Supplier: TERRAGENE
CT40
77506-912EA 15.67 USD
77506-912
Cintape® Self-Adhesive Indicator Tape for Hydrogen Peroxide (FDA)
Sterilization Indicators
Indicator tape for hydrogen peroxide sterilization processes.
  • Size: 50 m × 18 mm
  • Single-use chemical indicators that consist of a roll of self-adhesive plastic tape printed with indicator ink
  • These indicators have been designed to monitor Plasma or Vaporized Hydrogen Peroxide sterilization processes, ensuring an adequate exposure to the sterilizing agent during the sterilization process and allowing to distinguish between processed and unprocessed items
  • The adhesive component of the tape allows the adhesion to different types of packaging and wraps, such
More Product Information