DuraFrame® Rigid PCR Plates
Supplier: SCIENTIFIC SPECIALTIES, INC.
3730-00S 3730T00S 3735-00S 3735-01S 3735-02S 3735-04S 3735-05S 3735-06S 3735-09S 3735T00S 3742-00S 3742-01S 3742-02S 3742-04S 3742-05S 3742-06S 3742-09S 3742T00S 3750-00S 3750-01S 3750-02S 3750-04S 3750-05S 3750-06S 3750-09S
77506-814CS 525.3 USD
77506-814 77506-816 77506-818 77506-820 77506-822 77506-824 77506-826 77506-828 77506-830 77506-832 77506-838 77506-840 77506-842 77506-844 77506-846 77506-848 77506-850 77506-852 77506-858 77506-860 77506-862 77506-864 77506-866 77506-868 77506-870
DuraFrame® Rigid PCR Plates
Microplates PCR Plates
DuraFrame® plates feature a rugged polycarbonate frame, providing a solid and flat surface that guarantees seamless interaction with automated liquid handling platforms. With a firm grip assured, robots can confidently place DuraFrame® PCR plates with the utmost precision, eliminating any concerns of misalignment.
  • Polycarbonate frame is rigid and flat to facilitate a firm grip by robotic arms and ensure reliable placement
  • Clear polypropylene wells provide optimal thermal transfer and higher reaction efficiency
  • White well options enhance the fluorescence
More Product Information