Syringes for Syringe Pumps, XP
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2607715 2607752 2607752-BL 2607151 2607515 2607615 9607074 9607715 9607811 9607911 2647016 2647017 2647018 2647037 2647038 2647076 2647077 2647078 2647517 2647617 2607015 2607025 2607026 2607035 2607045 2607815 2607915 2647618 2647717 2647718 2647817 2647818 2647917 2647918 2647919 9607011 9607034 2607075 2607089
77506-242EA 76.25 USD
77506-242 77506-248 77506-634 77506-216 77506-222 77506-230 77506-622 77506-624 77506-626 77506-628 77506-430 77506-432 77506-434 77506-436 77506-438 77506-440 77506-442 77506-444 77506-446 77506-448 77506-162 77506-168 77506-170 77506-180 77506-186 77506-256 77506-268 77506-450 77506-452 77506-454 77506-456 77506-458 77506-460 77506-462 77506-464 77506-614 77506-620 77506-204 77506-208
Syringes for Syringe Pumps, XP
Syringes General-Purpose Syringes
XP series syringes are designed to work with most industry standard syringe pump designs.
  • Designed for volumes of 25 µl to 12.5 ml
  • Luer or ¹/₄ to 28 threaded end caps
  • Glass barrel: Borosilicate 3.3
  • Broad variety of syringe interfaces
  • Seals available in PTFE, UHMWPE, or custom UHMWPE blends
  • Customized designs and materials to suit application requirements
  • 3 cm stroke
  • A high chemical resistance makes this syringe a perfect heavy-duty syringes
PTFE Seal:
Chemically resistant
Heavy duty syringes
PTFE Black Seals
Specially designed for handling of strongMore Product Information