Heratherm™ Environmental Chambers
Supplier: Thermo Fisher Scientific
ECG750 ECS750 EIG750 EIS750 ESG750 ESS750
77486-596EA 26885 USD
77486-596 77486-598 77486-600 77486-602 77486-608 77486-610
Heratherm™ Environmental Chambers
Testing Chambers Environmental Test Chambers
Heratherm Environmental chambers offer exceptional environmental control in a hassle-free chamber that is easy to program and use.
  • Reduce your environmental footprint while achieving outstanding results with Heratherm Environmental chamber's decreased water and energy consumption and Peltier technology
  • Water-efficient humidification system with integrated water tank and optional clean water connection
  • Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors
  • Vertical low-energy LED lighting
More Product Information