Cubis® II Semi-Micro Balances
Supplier: Sartorius
MCA225P3S00DQP99ION MCA225P3S00DQP99MDS MCA225S3S00D MCA225S3S00DHWL MCA225S3S00DION MCA225S3S00DMDS MCA225S3S00DQP99 MCA225S3S00DQP99ION MCA225S3S00DQP99MDS MCA226S3S00D MCE225P3S00DHWL MCE225P3S00DION MCE225P3S00DMDS MCE225S3S00D MCE225S3S00DION MCE225S3S00DMDS MCA226S3S00DHWL MCA226S3S00DION MCA226S3S00DMDS MCA226S3S00DQP99 MCA226S3S00DQP99ION MCA226S3S00DQP99MDS MCE125S3S00D MCE125S3S00DION MCE125S3S00DMDS MCE225P3S00D MCA125S3S00D MCA125S3S00DHWL MCA125S3S00DION MCA125S3S00DMDS MCA225P3S00D MCA225P3S00DHWL MCA225P3S00DION MCA225P3S00DMDS MCA225P3S00DQP99 MCA225P3S00DQP99HWL
77476-508EA 21012.2 USD
77476-508 77476-510 77476-512 77476-514 77476-516 77476-518 77476-520 77476-522 77476-524 77476-526 77476-548 77476-550 77476-552 77476-554 77476-556 77476-558 77476-528 77476-530 77476-532 77476-534 77476-536 77476-538 77476-540 77476-542 77476-544 77476-546 77476-488 77476-490 77476-492 77476-494 77476-496 77476-498 77476-500 77476-502 77476-504 77476-506
Cubis® II Semi-Micro Balances
Balances and Scales
Semi-micro balances come with a readability of 0.01 mg or 10 µg and a maximum weighing capacity of up to 220 g.
  • Superior weighing performance: Fast measurement time and accurate weighing results are guaranteed thanks to next generation monolithic weighing system and the engineering solution compensating for environmental changes
  • No more issues with charged samples: Glass draft shields coated with conductive layer prevent outer electrostatic effects and the built-in ionizer effectively eliminates electrostatic charges from samples
  • Cleaning process guidance:
More Product Information