Cy3.5 tetrazine
Supplier: AAT BIOQUEST INC
903
77463-124EA 235.39 USD
77463-124
Cy3.5 tetrazine
Cy3.5 tetrazine
Cy3.5 tetrazine is an excellent building block that can be readily used to label TCO-modified biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only