BRAND® PCR Plates, 96-Well, Rigid Frame, BIO-CERT® PCR QUALITY
Supplier: Brandtech
781549 781541 781542 781543 781544 781545 781546 781547 781548
77447-876PK 242.24 USD
77447-876 77447-860 77447-862 77447-864 77447-866 77447-868 77447-870 77447-872 77447-874
BRAND® PCR Plates, 96-Well, Rigid Frame, BIO-CERT® PCR QUALITY
Microplates PCR Plates
BRAND® 96-well plates provide efficient PCR and qPCR solutions for medium to high sample throughput, featuring a strong polycarbonate frame and thin-walled wells for rapid temperature transfer.
  • Compatible with all standard thermal cyclers and fully automation ready
  • Select from semi-skirted or fully skirted plate options
  • Distinguish workflow steps easily with a range of color choices
  • White plates for qPCR, optimized to reflect fluorescence signals
BRAND® rigid frame PCR plates feature a robust polycarbonate frame, ensuring safe handling in both semi-More Product Information