Accessories for Compresstome® Legacy Specimen Tube Kits
Supplier: PRECISIONARY INSTRUMENTS LLC
VFBHSKVM12.5NC VFBHSKVM15.5NC VFSPSKVM12.5NC VFSPSKVM15.5NC
77435-404EA 1028.46 USD
77435-404 77435-406 77435-486 77435-488
Accessories for Compresstome® Legacy Specimen Tube Kits
Microtomes and Microtome Assemblies Microtomes
Compresstome 12.5 or 15.5 mm diameter legacy specimen tube kits, with or without blade holder.
  • Compatible with compresstome models VF-200, VF-200-0Z, VF-300, or VF-300-0Z
Tube kits with blade holder include 2 specimen tubes (12.5 or 15.5 mm) with matching plungers, 1 chilling block, 1 embedding ring, 1 tube holder/adaptor base for buffer tank, 1 set of screws/washers, 1 blade holder, 1 small Allen wrench, and 1 small screw.
Packaging: Tube kits without blade holder include 2 specimen tubes (12.5 or 15.5mm), one chilling block, one embedding ring,More Product Information