Accessories for Rotary Microtome RF-1000 Side Panel
Supplier: PRECISIONARY INSTRUMENTS LLC
RF1000SIDEBOX
77435-310EA 570.29 USD
77435-310
Accessories for Rotary Microtome RF-1000 Side Panel
Microtomes and Microtome Assemblies Microtomes
A replacement part for the external control panel on the RF-1000 rotary microtome.
  • Includes side control panel and cable
Control panel allows for control of slice thickness adjustments, movement of the specimen head, and start/stop of automated cutting.