1-bromo-2-fluoro-4-iodo-5-(trifluoromethoxy)benzene

Supplier: APOLLO SCIENTIFIC
Warning

PC99603-1G
77434-664EA 496.16 USD
77434-664
1-bromo-2-fluoro-4-iodo-5-(trifluoromethoxy)benzene
1-bromo-2-fluoro-4-iodo-5-(trifluoromethoxy)benzene
Formula: C₇H₂BrF₄IO
CAS Number: 2384187-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR