tert-Butanol 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

OR70032-1L
77410-970EA 95.76 USD
77410-970
tert-Butanol 99%
tert-Butanol
Formula: (CH₃)₃COH
Boiling Pt: 83 °C
Melting Pt: 23…26 °C
Density: 0.78 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR