Bromobenzene-[D₅] (99.5% D)

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Phenylbromide-D5, Bromobenzene-[²H₅]

DE188-5ML
77401-182EA 58.08 USD
77401-182
Bromobenzene-[D₅] (99.5% D)
Bromobenzene-[D5]
Formula: C₆D₅Br
MW: 161.97
Boiling Pt: 156.6
Melting Pt: -31
Density: 1.539 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000056
CAS Number: 4165-57-5
EINECS: 224-013-8
UN: 2514
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR