(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

A819521-100G A819521-10G A819521-250MG A819521-25G A819521-5G
103841-124EA 18 USD
103841-124 77293-874 77293-876 77293-878 77308-380 77308-382
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Caution: for Research Use Only

Formula: C₄₄H₃₂P₂
Melting Pt: 239…242 °C
Flash Pt: 419 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR