Maleimide 99%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrrole-2,5-dione

A518248-100G A518248-10G A518248-500G A518248-25G
103839-590EA 7 USD
103839-590 76818-848 76818-850 76818-852 76843-594 77307-700
Maleimide 99%
Maleimide

Caution: for Research Use Only

Formula: C₄H₃NO₂
Boiling Pt: 244 °C (760 mm Hg)
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005494
CAS Number: 541-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR