exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione 95%

Supplier: Ambeed
Warning

Synonyms: Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione, rel-(3aR,4S,7R,7aS)-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione, exo-2,3-Norbornanedicarboximide

A184020-100G A184020-10G A184020-1G A184020-25G A184020-500G
103815-700EA 5 USD
103815-700 103846-260 77029-560 77029-562 77301-876 77301-878 77871-914
exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione 95%
exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione

Caution: for Research Use Only

Formula: C₉H₁₁NO₂
Boiling Pt: 519.7 °C (760 mm Hg)
Melting Pt: 173…176 °C
Flash Pt: 171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12964181
CAS Number: 14805-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR