Atlanta Biomedical Syringe Infusion Pump, FDA Approved
Supplier: WILBURN MEDICAL, INC.
ABC-4100
77296-298EA 6168.56 USD
77296-298
Atlanta Biomedical Syringe Infusion Pump, FDA Approved
Pumps Syringe Pumps
The Atlanta BioMedical Corporation 4100 Syringe Infusion Pump recognizes most IV syringe manufacturers and accepts sizes from 1 ml to 120 ml. US made. Metal housing for durability. Includes a handle and can be mounted on any pole/rail system with an optional pole clamp.
  • Front syringe loading
  • Adjustable flow ranges – changes can be made without interrupting flow
  • Fast bolus rate storage for user defined settings
  • Multiple sensors to monitor delivery accuracy
  • AC or battery operation with 12 hours battery capacity
  • Full alarm capabilities
  • RS232 interface
More Product Information