Pegboards with 36-Piece DuraHook® Assortment, High Density Fiberboard
Supplier: Triton Products
PEG36-BLK PEG36-WHT
77269-754EA 87.43 USD
77269-754 77269-756
Pegboards with 36-Piece DuraHook® Assortment, High Density Fiberboard
Furniture Backsplashes, Wallracks, Wallpanels
These pegboards are made of high-grade HDF to ensure durability and are 4 times stronger than conventional pegboard.
  • Hole diameter is ¹/₄" with 1" O.C. spacing
  • Superior holding capacity when compared to standard pegboard
  • Abrasion, humidity and temperature resistant
  • Can be cut (using an 80-tooth or higher saw blade), drilled, or routed without cracking and fraying the board
  • Modulus of Rupture (MOR) rating of 5000 psi
  • Recommended to be used with our DuraHook® 70000 series locking pegboard hooks and accessories
HDF Board: High Density Fiberboard ¹/₄"More Product Information