18-Gauge Steel Square Hole Pegboard Strip with Mounting Hardware, 36" Width
Supplier: Triton Products
LBS-1S LBS-1W
77269-720EA 31.16 USD
77269-720 77269-722
18-Gauge Steel Square Hole Pegboard Strip with Mounting Hardware, 36" Width
Furniture Backsplashes, Wallracks, Wallpanels
LocBoard® is made from 18 gauge steel and offers the ultimate holding capacity for the most demanding tool and storage needs. It is 12 times stronger than conventional pegboard.
  • Multi-use strips can store almost any tool securely
  • All hooks lock securely on the strip and are easily repositioned in seconds with no damage to the pegboard
  • Mounting hardware is hidden from sight, presenting a clean, attractive, and durable appearance
  • 75 lbs. load rating
LocBoard® heavy-duty silver epoxy coated steel pegboard strips with square holes offers an ideal solutionMore Product Information