Pegboards with LocHook® Assortment, 18-Gauge Steel Square Hole
Supplier: Triton Products
LB18-1BH-KIT LB18-1GH-KIT LB18-1WH-KIT
77269-636EA 106.94 USD
77269-636 77269-638 77269-640
Pegboards with LocHook® Assortment, 18-Gauge Steel Square Hole
Furniture Backsplashes, Wallracks, Wallpanels
LocBoard® is made from 18 gauge steel and offers the ultimate holding capacity for the most demanding tool and storage needs. It is 12 times stronger than conventional pegboard.
  • Ideal for heavy, oversized and odd shaped professional power tools, hand, maintenance tools and more
  • Use with LocHook® 50000 series 100% heavy-duty steel two and four point contact locking hooks
  • All hooks lock securely on the board and are easily repositioned
  • Mounting hardware is hidden from sight - presenting a clean, attractive and durable appearance
  • 350 lbs. load rating per
More Product Information