DuraBoard® Pegboards, Black ABS Textured
Supplier: Triton Products
DB-4BK DB-96BLK DB-2BK
77269-614PK 409.63 USD
77269-614 77269-622 77269-602
DuraBoard® Pegboards, Black ABS Textured
Furniture Backsplashes, Wallracks, Wallpanels
DuraBoard® is the most durable, weather and impact resistant pegboard available. It is eight times stronger than conventional pegboard.
  • Hole diameter is ¹/₄" with 1" O.C spacing
  • DuraBoard will never fade, dry out, crack or peel over time so it maintains holding capability over many years of use
  • Offers a professional looking pegboard system that can be cleaned without harming the board
  • Board surface is ideal for creating silhouettes of tools to be stored and managed allowing for easy tool identification
  • Use Triton 70016 mounting spacer kit to mount
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable