VWR® Bagged Examination Gloves, Nitrile
Supplier: VWR International
77257-070 77257-068 77257-066 77257-072 77257-062 77257-060 77257-058 77257-064
77257-070CS 220 USD
77257-070 77257-068 77257-066 77257-072 77257-062 77257-060 77257-058 77257-064
VWR® Bagged Examination Gloves, Nitrile
Gloves
VWR® bagged examination gloves provide quality and comfort. Perfect for cleanroom, laboratory, and ESD environments.
  • Bagged gloves for cleanroom compatibility
  • Powder-free nitrile
  • Cleanroom class 10,000 (DI washed)
  • Static control
  • Surface resistance: 10¹¹ Ω
  • Volume resistance: 10⁹ Ω
VWR® bagged examination gloves are powder-free nitrile, cleanroom compatable gloves that offer static control and provide a high level of quality and comfort. These gloves are available in a blue or white finish.
Certifications: CAFE RGP: 10⁹ ohms as per ANSI/ESD SP15.1.
More Product Information