2,2'-Dihydroxybiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Biphenol, 2,2-Biphenol, Biphenyl-2,2'-diol

A248618-1000G A248618-100G A248618-25G A248618-500G
77249-932EA 87 USD
77249-932 77249-934 77249-936 77249-938
2,2'-Dihydroxybiphenyl 98%
2,2'-Dihydroxybiphenyl

Caution: for Research Use Only

Formula: C₁₂H₁₀O₂
MDL Number: MFCD00002210
CAS Number: 1806-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR