Pyruvic acid 96%

Supplier: Ambeed
Danger

Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid

A765171-10G A765171-25G A765171-100G
77158-978EA 5 USD
77158-978 77158-980 77686-777
Pyruvic acid 96%
Pyruvic acid

Caution: for Research Use Only

Formula: C₃H₄O₃
MDL Number: MFCD00002585
CAS Number: 127-17-3
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR