Fmoc-D-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Tyr-OH, N-Fmoc-D-tyrosine

A629999-100G A629999-1G A629999-25G A629999-5G
77157-844EA 327 USD
77157-844 77157-846 77157-848 77157-850
Fmoc-D-tyrosine 98%
Fmoc-D-tyrosine

Caution: for Research Use Only

Formula: C₂₄H₂₁NO₅
MW: 403.434 g/mol
MDL Number: MFCD00171384
CAS Number: 112883-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR