Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate 98%

Supplier: Ambeed
Warning

Synonyms: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate

A471068-100G A471068-25G A471068-50MG A471068-5G A471068-1G
77156-326EA 46 USD
77156-326 77156-328 77156-330 77156-332 77686-253
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate 98%
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate

Caution: for Research Use Only

Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MDL Number: MFCD00150755
CAS Number: 34369-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR