Service for Advanced MS and ML Balances
Supplier: Mettler Toledo
B99504130101 B99504130102 B99484130101 B99484130102 B99494130101 B99494130102 B99474130101 B99474130102 G17044130101 G17044130102
77135-240EA 1562.87 USD
77135-240 77135-242 77135-204 77135-206 77135-224 77135-226 77135-184 77135-186 77322-204 77322-206
Service for Advanced MS and ML Balances
Services
Professional installation completed by a trained Mettler Toledo service technician will guarantee maximum performance and optimal use.
  • Achieve peak performance and compliance with quality systems and regulatory requirements
  • Take the right steps at the beginning of equipment use
  • The Accuracy Calibration Certificate (ACC) is the new benchmark in the calibration of weighing equipment
  • Tests include repeatability, eccentricity and error of indication
  • A statement of measurement uncertainty is also included
StarterPac includes: Professional installationMore Product Information