Seals, Optimize Technologies
Supplier: Avantor
31-36-00806 34-59-00902 34-59-00907 26-59-00647 26-36-00633 26-36-00635 39-69-01262 39-69-01267 22-67-00516 22-68-00516 22-68-00520 22-67-00520 34-36-00863 34-36-00864 34-36-00865 26-36-00630 26-36-00631 26-36-00632 24-36-00573 24-36-00575 32-36-01303 32-36-01304 35-36-00938 35-36-00939 39-31-01147 39-30-01124 38-36-01002 38-36-01003 38-59-01038 38-59-01043 33-36-00804 33-36-00805 25-59-00613 25-36-00597 25-36-00598 25-59-00612 24-36-00574 22-69-00516 22-69-00520 24-34-00559 24-46-00585 24-46-00589 24-47-00585 24-48-00585 24-48-00589 24-49-00585 24-49-00589 24-56-00576 24-56-00581 24-57-00576 24-57-00581 24-58-00576 24-58-00581 24-59-00576 24-59-00581 25-36-00599 25-36-00600 25-36-00601 25-36-00602 25-58-00612 25-58-00613 26-56-00638 26-56-00647 26-57-00638 26-57-00647 26-58-00638 26-58-00647 26-59-00638 27-34-00658 27-36-01570 27-36-01571 27-36-01572 27-36-01573 27-36-02364 27-36-02365 27-36-02366 27-36-02367 28-36-00690 28-36-00691 28-36-00693 28-46-02092 28-46-02094 28-47-02092 28-47-02094 28-48-02092 28-48-02094 28-49-02092 28-49-02094 31-34-00738 31-36-00807 31-56-00759 31-56-00764 31-57-00759 31-57-00764 31-58-00759 31-58-00764 31-59-00759 31-59-00764 32-34-00771 32-46-00782 32-46-00787 32-47-00782 32-47-00787 32-48-00782 32-48-00787 32-49-00782 32-49-00787 33-36-00806 33-36-00807 33-56-00830 33-56-00835 33-57-00830 33-57-00835 33-58-00830 33-58-00835 33-59-00830 33-59-00835 34-34-01345 34-36-00866 34-46-00885 34-46-00893 34-47-00885 34-47-00893 34-48-00885 34-48-00893 34-49-00885 34-49-00893 34-56-00902 34-56-00907 34-57-00902 34-57-00907 34-58-00902 34-58-00907 35-32-01333 35-36-00916 35-36-00917 35-36-00920 35-36-00921 35-36-00930 35-36-00931 35-56-00952 35-56-00956 35-56-00961 35-57-00952 35-57-00956 35-57-00961 35-58-00952 35-58-00956 35-58-00961 35-59-00952 35-59-00956 35-59-00961 38-34-00999 38-36-01000 38-36-01001 38-36-01553 38-36-01554 38-56-01038 38-56-01043 38-56-01067 38-56-01544 38-56-01546 38-57-01038 38-57-01043 38-57-01067 38-57-01544 38-57-01546 38-58-01038 38-58-01043 38-58-01067 38-58-01544 38-58-01546 38-59-01067 38-59-01544 38-59-01546 39-24-01088 39-24-01090 39-24-01091 39-24-01290 39-24-01291 39-30-01113 39-31-01151 39-31-01171 39-34-01191 39-34-02180 39-36-01206 39-36-01216 39-36-01217 39-36-01305 39-36-01306 39-36-02124 39-36-02127 39-36-02228 39-36-02229 39-36-02596 39-36-02597 39-46-01949 39-47-01949 39-47-01950 39-48-01907 39-48-01909 39-48-01949 39-48-01950 39-49-01907 39-49-01909 39-49-01949 39-49-01950 39-56-01251 39-56-01256 39-57-01251 39-57-01256 39-58-01251 39-58-01256 39-59-01251 39-59-01256 39-66-01262 39-66-01267 39-67-01262 39-67-01267 39-68-01262 39-68-01267 42-34-01354 42-36-01355 42-36-01356 42-36-01357 42-36-01358 42-56-01038 42-56-01043 42-56-01544 42-56-01546 42-57-01038 42-57-01043 42-57-01544 42-57-01546 42-58-01038 42-58-01043 42-58-01544 42-58-01546 42-59-01038 42-59-01043 42-59-01544 42-59-01546 44-36-00437 44-36-00439 39-36-02855 39-36-02856 39-36-02860 39-38-01308 39-38-01311 39-36-01214 39-36-01215 22-38-01311 28-36-00692 34-47-02644 42-32-02841 34-46-02158 34-46-02627 39-46-02168 39-66-02169 39-48-02170 24-46-02141 31-56-02152 34-34-02650 39-36-01207 25-32-02820 27-31-02853 27-32-02844 34-48-02644 39-32-02825 39-36-02603 28-46-02148 35-56-02162 39-56-02166 39-66-02167 39-46-02187 24-56-02140 25-58-02143 26-56-02144 42-56-02151 38-56-02164 38-56-02165 34-56-02159 38-56-01704 33-56-02157 39-34-01188 25-32-02081 27-32-02845 27-36-02851 34-46-02644 34-49-02644 39-32-02823 35-56-02161 39-56-02186 42-56-02150 31-56-02153 27-04-02852 25-34-04543
21010-580EA 311.99 USD
21010-580 21509-805 21509-826 21509-828 21509-866 21509-872 21509-888 21509-899 21509-901 21509-903 21509-905 21509-906 21509-908 21509-962 21509-964 21509-967 21510-006 21510-008 21510-010 21510-034 21510-036 21510-040 21510-042 21510-050 21510-052 21510-269 21510-294 21510-370 21510-372 21510-410 21510-412 21510-422 21510-424 21510-438 21510-440 21510-448 21510-460 21510-462 21510-538 21510-580 77100-540 77100-542 77100-576 77100-582 77100-584 77100-586 77100-590 77100-592 77100-594 77100-596 77100-598 77100-600 77100-602 77100-604 77100-606 77100-608 77100-610 77100-612 77100-618 77100-620 77100-622 77100-624 77100-628 77100-630 77100-644 77100-646 77100-648 77100-650 77100-652 77100-654 77100-656 77100-664 77100-672 77100-674 77100-676 77100-678 77100-680 77100-684 77100-686 77100-690 77100-712 77100-714 77100-716 77100-722 77100-724 77100-730 77100-732 77100-738 77100-740 77100-746 77100-748 77100-780 77100-784 77100-786 77100-788 77100-790 77100-792 77100-794 77100-796 77100-798 77100-800 77100-802 77100-808 77100-814 77100-816 77100-818 77100-820 77100-822 77100-824 77100-826 77100-828 77100-864 77100-866 77100-868 77100-870 77100-874 77100-876 77100-882 77100-884 77100-890 77100-892 77100-898 77100-900 77100-924 77100-930 77100-938 77100-946 77100-948 77100-950 77100-952 77100-954 77100-956 77100-958 77100-960 77100-962 77100-964 77100-966 77100-968 77100-970 77100-972 77100-974 77100-976 77100-978 77100-980 77100-982 77100-984 77100-986 77100-988 77100-998 77101-002 77101-004 77101-006 77101-008 77101-010 77101-012 77101-014 77101-016 77101-018 77101-026 77101-028 77101-030 77101-032 77101-034 77101-036 77101-038 77101-040 77101-042 77101-044 77101-046 77101-048 77101-098 77101-100 77101-102 77101-104 77101-106 77101-108 77101-110 77101-112 77101-114 77101-116 77101-118 77101-120 77101-122 77101-124 77101-126 77101-128 77101-130 77101-132 77101-134 77101-136 77101-138 77101-140 77101-142 77101-144 77101-146 77101-148 77101-164 77101-166 77101-168 77101-170 77101-172 77101-192 77101-210 77101-218 77101-238 77101-246 77101-250 77101-254 77101-256 77101-258 77101-260 77101-262 77101-264 77101-266 77101-268 77101-270 77101-272 77101-294 77101-302 77101-304 77101-310 77101-312 77101-314 77101-316 77101-322 77101-324 77101-326 77101-328 77101-338 77101-340 77101-346 77101-348 77101-354 77101-356 77101-362 77101-364 77101-366 77101-368 77101-370 77101-372 77101-374 77101-376 77101-412 77101-414 77101-416 77101-418 77101-420 77101-428 77101-430 77101-432 77101-434 77101-436 77101-438 77101-440 77101-442 77101-444 77101-446 77101-448 77101-450 77101-452 77101-454 77101-456 77101-458 77101-466 77101-468 87006-036 87006-038 87006-042 21509-068 21509-102 21509-134 21509-136 21509-187 21509-248 46600-650 83008-352 83008-386 83008-424 83008-426 83008-440 83008-444 83008-446 83008-460 83008-464 83008-466 83008-472 83008-490 21010-582 46600-640 83008-326 83008-342 83008-348 83008-388 83008-412 83008-418 83008-438 83008-450 83008-454 83008-458 83008-462 83008-470 83008-476 83008-478 83008-486 83008-506 83008-508 83008-442 83008-452 83008-500 46600-642 83008-324 83008-350 83008-354 83008-384 83008-390 83008-410 83008-448 83008-456 83008-484 83008-492 89522-328 89522-404
Seals, Optimize Technologies
Seals Mechanical Seals
Piston seals are designed to offer reliable performance in a variety of operating conditions.
  • Polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used
  • Graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern
  • Reliable performance in a variety of operating conditions
Installing a piston seal can be a tricky undertaking. A relatively small seal must be fit into the seal cavity in such a way to ensure thatMore Product Information