OPTI-LOK™ Low Pressure Fittings
Supplier: Optimize
10-21-00300 10-21-00317 10-21-00318 10-21-00284 10-21-00293 10-21-00296 10-21-00306 10-21-00308 10-21-00310 10-21-00343 10-21-00281 10-21-00283 10-21-00290 10-21-00291 10-21-00295 10-21-00299 10-21-00305 10-21-00307 10-21-00312 10-21-00313 10-21-00323 10-21-00326 10-22-00315 10-22-00319
21510-187EA 13.91 USD
21510-187 21510-199 21510-201 21510-702 21510-712 21510-714 21510-718 21510-720 21510-722 21510-776 77100-192 77100-194 77100-198 77100-200 77100-206 77100-210 77100-214 77100-216 77100-222 77100-224 77100-230 77100-232 77100-246 77100-248
OPTI-LOK™ Low Pressure Fittings
Fittings and Connectors
OPTI-LOK LP Fittings are an excellent choice for making low-pressure connections into ¹/₄-28 ports. Connections to flat-bottomed ports can be made using OPTI-LOK flat-bottomed ferrules for ¹/₁₆ or ¹/₈" tubing without the need for pre-flanged tubing ends. OPTI-LOK LP Fittings are precision machined to deliver leak-free, zero-dead-volume connections every time. We also offer ¹/₄-28 unions in various through-hole sizes for joining flat-bottom fittings.
  • PEEK, ETFE, PTFE, PCTFE, black and white acetal nuts and ferrules for ¹/₈", 0.100" and ¹/₁₆" outer Ø tubing
More Product Information