6-Amino-5-bromonicotinonitrile 97%

Supplier: Ambeed
Warning

A123490-1G A123490-250MG A123490-5G A123490-10G A123490-25G
77026-614EA 14.86 USD
77026-614 77026-616 77026-618 77026-620 77871-551 77871-552
6-Amino-5-bromonicotinonitrile 97%
6-Amino-5-bromonicotinonitrile

Caution: for Research Use Only

Formula: C₆H₄BrN₃
MDL Number: MFCD02102412
CAS Number: 477871-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR