PTS Panels® CHOL + GLU Test Strips
Supplier: PTS Diagnostics
1765
77020-346EA 182.03 USD
77020-346
PTS Panels® CHOL + GLU Test Strips
Clinical Diagnostic Supplies Clinical Reagents and Kits
PTS Panels CHOL+GLU test strips provide lab-quality total cholesterol and glucose results on-site in about 45 seconds from a single fingerstick.
  • Measures total cholesterol and glucose
  • 25 test strips per vial
  • Room temperature storage
PTS Panels CHOL+GLU test strips are for professional use with CardioChek PA and CardioChek Plus analyzers to measure total cholesterol and glucose in venous whole blood or capillary fingerstick blood.
This test system will provide numeric results in the following ranges:
Cholesterol: 100 to 400 mg/dl (2.59 to 10.36 mmol/L)
Glucose: 20 to 600 mg/dl (1.11 to 33.3 mmol/L)