Lighthouse Pre-Mix Electric Foam Unit
Supplier: Unimed-Midwest
FPCF516IJR
77007-804EA 1702.55 USD
77007-804
Lighthouse Pre-Mix Electric Foam Unit
Spray Guns
This 120 VAC electric unit lets users apply disinfectants and other chemistry with a handheld wand. With a built-in air compressor, simply plug the unit into a standard wall outlet and get to work.
  • Draws from pre-mixed solution
  • Powered by electricity (120 VAC)
  • Built-in air compressor
  • 15 micron droplet size at 50 psi (3.4 bar)
  • Compact size for portability
  • Santoprene, Viton, or Kalrez pump seals for chemical compatibility
Ideal for fogging/misting in areas without compressed air, this unit features a handheld wand with 50' (15.2 m) of twin-lineMore Product Information