BX-795 98%

Supplier: Ambeed
Warning

Synonyms: BX795, N-(3-((5-Iodo-4-((3-(thiophene-2-carboxamido)propyl)amino)pyrimidin-2-yl)amino)phenyl)pyrrolidine-1-carboxamide

A700044-10MG A700044-1MG A700044-25MG A700044-50MG A700044-5MG A700044-100MG A700044-250MG
76981-086EA 51 USD
76981-086 76981-088 76981-090 76981-092 76981-094 77673-914 77673-916
BX-795 98%
BX-795

Caution: for Research Use Only

Formula: C₂₃H₂₆IN₇O₂S
MDL Number: MFCD12546134
CAS Number: 702675-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR