5-Amino-2-chloro-3-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-chloro-3-picoline, 6-Chloro-5-methylpyridin-3-amine

A127047-100G A127047-10G A127047-1G A127047-25G A127047-5G
76971-908EA 171 USD
76971-908 76971-910 76971-912 76971-914 77670-792
5-Amino-2-chloro-3-methylpyridine 97%
5-Amino-2-chloro-3-methylpyridine

Caution: for Research Use Only

Formula: C₆H₇ClN₂
MDL Number: MFCD03095087
CAS Number: 38186-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR