VWR® Deep Well Plates, 96-Well and 384-Well
Supplier: VWR International
76924-064 76924-066 76924-044 76924-046 76924-048 76924-050 76924-056 76924-058
76924-064CS 287.82 USD
76924-064 76924-066 76924-044 76924-046 76924-048 76924-050 76924-056 76924-058
VWR® Deep Well Plates, 96-Well and 384-Well
Microplates Deep Well Microplates
VWR® DeepWell plates are ideal for sample collection, preparation, and long-term storage.
  • Centrifugation to 4000×g
  • Manufactured from virgin polypropylene to ensure purity and sample integrity, chemical resistance, and compatibility with long-term storage needs
  • Alphanumeric indexing molded in on A-H and 1-12 axes
  • Optimum sample mixing and recovery with highly-polished wells and radiused corners
  • Raised rims on each well facilitate secure sealing with films and sealing mats
  • Bar coded plates available—please inquire
VWR® 96- and 384-well deep well platesMore Product Information