Avantor® Magnetic Mixing System
Supplier: Avantor Fluid Handling
AFH0200MMS-01004 AFH0500MMS-01004
76894-046EA 60001.62 USD
76894-046 76894-056
Avantor® Magnetic Mixing System
Stirrers Overhead Stirrers
The Avantor Magnetic Mixing System is a robust, flexible, high-quality, expert-designed solution for single use mixing needs.
  • Intuitive software: Controlled by a user-friendly software program that runs on a 7" color touchscreen display. This software allows users to save mixing profiles and customize their mixing settings
  • Integrated loadcell: The system can have an integrated loadcell option that measures the weight of the vessel and contents, providing accurate feedback on the mixing process
  • Temperature controlled jacket: The system has a temperature-
More Product Information