2-Amino-5-(trifluoromethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-amino-5-(trifluoromethoxy)benzoic acid

A377097-1G A377097-250MG A377097-25G A377097-5G
76876-606EA 11 USD
76876-606 76876-608 76876-610 76876-612
2-Amino-5-(trifluoromethoxy)benzoic acid 97%
2-Amino-5-(trifluoromethoxy)benzoic acid

Caution: for Research Use Only

Formula: C₈H₆F₃NO₃
MDL Number: MFCD00176651
CAS Number: 83265-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR