Gas Chromatograph with FID (GC-FID)
Supplier: LUCIDITY SYSTEMS LLC
928270-120 NP-1397 928270-240
76836-910EA 19243.92 USD
76836-910 76836-958 77569-836
Gas Chromatograph with FID (GC-FID)
Chromatography Systems Gas Chromatographs
Compact, economical, fully functional gas chromatograph.
  • Lucidity quick column change for quick and easy column installation
  • Equipped with a split/splitless injection port
  • Contains a very small oven which allows for rapid heating and cooling
  • Intuitive Lucidity software included with purchase of instrument
  • Instrument changes color to indicate status
  • Self-installable and ease of use design
  • Low power consumption
The Lucidity GC has one of the smallest footprints of any fully functional GC-FID. Changing GC columns in the Lucidity GC is as simple asMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
W×D×H 10.5×18×11.5"