1,3-Dimethyl-2-imidazolidinone 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Dimethylimidazolidin-2-one

A490230-100G A490230-25G A490230-500G A490230-1000G
76830-070EA 20 USD
76830-070 76830-072 76830-074 77551-486
1,3-Dimethyl-2-imidazolidinone 98%
1,3-Dimethyl-2-imidazolidinone

Caution: for Research Use Only

Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Boiling Pt: 225.5
Melting Pt: 8
Flash Pt: 93
MDL Number: MFCD00003188
CAS Number: 80-73-9
Merck Index: 13,03275

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR