VWR® Cryogenic Dot and Strip Labels
Supplier: VWR International
76808-834 76808-874 76808-956 76808-898 76808-954 76808-932 76808-826 76808-946 76808-940 76808-876 76808-844 76808-924 76808-942 76808-974 76808-854 76808-978 76808-848 76808-896 76808-928 76808-926 76808-980 76808-882 76808-964 76808-858 76808-902 76808-938 76808-914 76808-884 76808-866 76808-916 76808-934 76808-894 76808-856 76808-846 76808-968 76808-930 76808-906 76808-970 76808-860 76808-950 76808-936 76808-976 76808-886 76808-892 76808-862 76808-868 76808-944 76808-904 76808-880 76808-908 76808-888 76808-836 76808-832 76808-910 76808-922 76808-838 76808-872 76808-890
76808-834RL 73.91 USD
76808-834 76808-874 76808-956 76808-898 76808-954 76808-932 76808-826 76808-946 76808-940 76808-876 76808-844 76808-924 76808-942 76808-974 76808-854 76808-978 76808-848 76808-896 76808-928 76808-926 76808-980 76808-882 76808-964 76808-858 76808-902 76808-938 76808-914 76808-884 76808-866 76808-916 76808-934 76808-894 76808-856 76808-846 76808-968 76808-930 76808-906 76808-970 76808-860 76808-950 76808-936 76808-976 76808-886 76808-892 76808-862 76808-868 76808-944 76808-904 76808-880 76808-908 76808-888 76808-836 76808-832 76808-910 76808-922 76808-838 76808-872 76808-890
VWR® Cryogenic Dot and Strip Labels
Labels
VWR® Cryogenic dot and strip labels are ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications.
  • Can be placed in most incubators
  • DYMO or Zebra compatible
  • Direct thermal and cryogenic
Labels feature a unique adhesive that can withstand extreme temperatures from –196 to 121 °C (–321 to 250 °F), without the need for lamination.
Ordering information: Rolls of labels are supplied on a 44 mm Ø (1.75") core. Laser printer label sheets are 21.6W×27.9L cm (8.5×11"). Assorted labels come in blue, green,More Product Information