DURAN® Bottle Carrying Systems
Supplier: DWK Life Sciences (KIMBLE)
292806304 292807309 292816305 292826306 292836307
76808-784CS 39.25 USD
76808-784 76808-792 76808-782 76808-788 76808-780 76808-794 76808-790
DURAN® Bottle Carrying Systems
Carriers Bottle Carriers/Containers
The ergonomic design offers health and safety benefits and allows easy and safe transport of bottles.
  • Increase comfort and safety when pouring liquids from larger bottles
  • Collar is only removable by cutting, making the attachment 100% secure in use
  • Carrier can be cleaned attached to the bottle in an automated washer
  • Autoclavable
The Bottle Carrying System range fits standard laboratory bottles that have a GL 45 neck finish, and are available for 2 or 5 L bottle sizes. The carriers are ergonomically designed and offer health and safety benefits to theMore Product Information