Brady® StainerBondz™ Microscope Slide Matte Polyester Labels
Supplier: Brady Worldwide
170884 170885 170886 175660
76795-846EA 104.28 USD
76795-846 76795-872 76795-886 76795-728
Brady® StainerBondz™ Microscope Slide Matte Polyester Labels
Labels
Specialized labels are available in pre-sized shapes for commonly used slides and tissue cassettes in which superior chemical resistance is required.
  • Permanent acrylic adhesive
  • Matte finish
  • Autoclavable material
  • Recommended ribbon: R6400
Durable labels withstand the rigors of typical deparaffinization, including ethanol and xylene, as well as hematoxylin and eosin slide staining processes. Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme conditions.
Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).
Order Now
SPECIFICATIONS
Environmentally Preferable