7-Deazaadenosine 98%

Supplier: Ambeed
Danger

Synonyms: 4-Amino-7-(β-D-ribofuranosyl)pyrrolo[2,3-d]pyrimidine

A118999-100MG A118999-10MG A118999-1G A118999-1MG A118999-250MG A118999-50MG A118999-5MG
76781-602EA 99 USD
76781-602 76781-604 76781-606 76781-608 76781-610 76781-612 76781-614
7-Deazaadenosine 98%
7-Deazaadenosine

Caution: for Research Use Only

Formula: C₁₁H₁₄N₄O₄
MDL Number: MFCD00056012
CAS Number: 69-33-0
UN: 3462
ADR: 6.1,Ⅱ

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR