α,α,α,α',α',α'-Hexafluoro-5-nitroxylene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Nitro-3,5-bis(trifluoromethyl)benzene

A510763-1G A510763-100G A510763-25G A510763-5G
76774-568EA 8 USD
76774-568 77551-532 77551-534 77602-192
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene 98%
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene

Caution: for Research Use Only

Formula: C₈H₃F₆NO₂
MDL Number: MFCD00000384
CAS Number: 328-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR