(2,3-Difluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,3-Difluorophenyl)acetic acid

A313966-25G A313966-5G A313966-100G
76772-824EA 34 USD
76772-824 76772-826 77877-999
(2,3-Difluorophenyl)acetic acid 98%
(2,3-Difluorophenyl)acetic acid

Caution: for Research Use Only

Formula: (C₆H₃F₂)CH₂COOH
MDL Number: MFCD00040968
CAS Number: 145689-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR