Diphenyl ether 99%

Supplier: Ambeed
Danger

Synonyms: Diphenyl oxide, Phenyl ether, Phenoxybenzene

A931916-25G A931916-500G A931916-100G A931916-1000G
76729-232EA 5 USD
76729-232 77552-728 77569-652 77573-860
Diphenyl ether 99%
Diphenyl ether

Caution: For research use only.

Formula: C₁₂H₁₀O
MDL Number: MFCD00003034
CAS Number: 101-84-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR