Silver sulfate 99%

Supplier: Ambeed
Danger

Synonyms: Disilver sulfate, Silver(I) sulfate, Disilver sulphate

A289249-10G A289249-25G A289249-5G A289249-500G
76723-464EA 31 USD
76723-464 76723-466 76723-468 77601-286
Silver sulfate 99%
Silver sulphate

Caution: for Research Use Only

Formula: Ag₂SO₄
MDL Number: MFCD00003407
CAS Number: 10294-26-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR