Cesium carbonate 98%

Supplier: Ambeed
Warning

A171817-100G A171817-10G A171817-500G A171817-1000G A171817-25G A171817-5G
103809-050EA 231 USD
103809-050 103844-946 76690-198 76690-200 76690-202 77082-120 77301-160 77301-162
Cesium carbonate 98%
Cesium carbonate

Caution: for Research Use Only

Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: 610
Flash Pt: 169.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
Merck Index: 13,02020

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR