Sol-Care® Heel Blood Lancets
Supplier: SOL-MILLENNIUM MEDICAL
LAH001 LAH002 LAH003 LAH004
76645-658BX 148.46 USD
76645-658 76645-660 76645-662 76645-664
Sol-Care® Heel Blood Lancets
Blood Lancets
The Sol-Care safety heel lancet is designed with a surgical grade blade, delivering a smooth, precise sweeping motion, that minimizes bruising and trauma during blood collection.
  • Ergonomic design offers multiple grip options
  • Easy-to-identify colors and size indicators on side of device (Preemie/Full Term) make choosing the correct heel-stick easy for the user
  • Incision mark for an accurate lance
Designed to quickly and comfortably sample neonatal blood with minimal pain and discomfort.
Certifications: Meets or exceeds OSHA and NCCLS guidelines forMore Product Information