Sterile Empty Vials, Assembled and Pyrogen Free
Supplier: ALK LIFE SCIENCE SOLUTIONS
SEV520G SEV520R SEV520S SEV520S-AMB SEV520Y SEV100 SEV100-AMB SEV100-FLUR SEV1020B SEV1020G SEV1020R SEV1020S SEV1020S-AMB SEV1020S-FLUR SEV1020Y SEV30 SEV30-AMB SEV30-FLUR SEV50 SEV50-AMB SEV50-FLUR SEV520B SEV20-16 SEV213-25 SEV213-25-AMB
76640-542PK 84.69 USD
76640-542 76640-580 76640-544 76640-582 76640-556 76640-538 76640-560 76640-562 76640-574 76640-564 76640-540 76640-546 76640-578 76640-548 76640-566 76640-558 76640-554 76640-552 76640-576 76640-568 76640-550 76640-570 76640-534 76640-572 76640-536 76994-118 76994-120 76994-122
Sterile Empty Vials, Assembled and Pyrogen Free
Vials
ALK Life Science Solutions provides high quality sterile empty vials to maintain the purity and integrity of what is inside.
  • Type 1 glass USP borosilicate
  • Latex free stopper
  • Produced at a cGMP FDA manufacturer
  • USP tested for sterility and endotoxins (pyrogen free)
  • Assembled in the USA, in an ISO 5 (class 100) cleanroom
Ideal for analytical laboratories for specimen containment and transfer. The sterilization processes for ALK Life Science solutions sterile empty vials are carried out under strict Class 100 U.S. Pharmacopoeia and Federal Code ofMore Product Information