Heat Sealing Foils, Peelable, DMSO Resistant
Supplier: AZENTA US, INC
4TI-0586-SLR 4TI-0587 4TI-0585 4TI-0585/S 4TI-0585-SLR 4TI-0586 4TI-0586/S
76637-694EA 934.21 USD
76637-694 76637-860 76637-726 76637-814 76637-766 76637-894 76637-890
Heat Sealing Foils, Peelable, DMSO Resistant
Seals Microplate Seals
The DMSO resistant peel heat seal is a foil seal compatible with polypropylene and forming an excellent seal to cyclic olefin copolymer (COC) plates.
  • Permanent seal to polyethylene
  • Peelable seal to polypropylene and COC
  • High solvent resistance, including 100% DMSO
  • Suitable for long term storage
  • For low and room temperature compound storage in Dimethyl Sulfoxide (DMSO) and organic solvents
  • Temperature range: –80 to +40 °C
  • Non sterile
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick andMore Product Information